missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Venlafaxine hydrochloride

Product Code. 18085882
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18085882 50 mg 50mg
18074192 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18085882 Supplier R&D Systems™ Supplier No. 2917/50
  • ONLINE EXCLUSIVE PRICE
    616.93 €
    556.50 € / 50mg
    Save 60.43 €
    9% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. 5-HT and NA reuptake inhibitor (SNRI)

Dual serotonin/noradrenalin re-uptake inhibitor that displays ∽ 30-fold higher affinity for SERT than NET (Ki values are 82 and 2480 nM respectively). Antidepressant; increases swimming and climbing behavior in the forced-swim test in rats.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99300-78-4
Molecular Formula C17H28ClNO2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 313.86
Purity >99%
Molecular Weight (g/mol) 313.866
Chemical Name or Material Venlafaxine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.