missing translation for 'onlineSavingsMsg'
Learn More

Venlafaxine hydrochloride, MedChemExpress

Product Code. 16468408
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468408 10 mg 10mg
16444297 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468408 Supplier Medchem Express Supplier No. HYB0196A
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Venlafaxine hydrochloride (Wy 45030 hydrochloride) is an orally active, potent serotonin (5-HT)/norepinephrine (NE) reuptake dual inhibitor. Venlafaxine is an antidepressant.

Venlafaxine hydrochloride (Wy 45030 hydrochloride) is an orally active, potent serotonin (5-HT)/norepinephrine (NE) reuptake dual inhibitor. Venlafaxine is an antidepressant.

Specifications

Chemical Name or Material Venlafaxine hydrochloride
CAS 99300-78-4
Color White
Purity Grade Notes Research
Molecular Formula C17H28ClNO2
Quantity 10 mg
Synonym Wy 45030 hydrochloride
Solubility Information H2O : ≥ 100 mg/mL (318.61 mM) ∣DMSO : 50 mg/mL (159.31 mM; Need ultrasonic)
SMILES OC1(C(C2=CC=C(OC)C=C2)CN(C)C)CCCCC1.Cl
Molecular Weight (g/mol) 313.86
Formula Weight 313.86
Percent Purity 99.86%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.