missing translation for 'onlineSavingsMsg'
Learn More

Verapamil-d3 Hydrochloride, TRC

Product Code. 30457282
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457282 5 mg 5mg
30451709 50 mg 50mg
30455563 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30457282 Supplier Toronto Research Chemicals Supplier No. TRCV1250015MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spezifikation

Chemical Name or Material Verapamil-d3 Hydrochloride
CAS 2714485-49-9
Molecular Formula C27 D3 H35 N2 O4 . Cl H
Quantity 5 mg
Synonym (±)-Verapamil-d3 hydrochloride, Verapamil-d3 hydrochloride, Verapamil-D3 (N-methyl-D3) hydrochloride, 5-[(3,4-Dimethoxyphenethyl)methyl-d3-amino]-2-(3,4-dimethoxyphenyl)-2-isopropylvaleronitrile hydrochloride, α-[3-[[2-(3,4-Dimethoxyphenyl)ethyl]methyl-d3-amino]propyl]-3,4-dimethoxy-α-(1-methylethyl)benzeneacetonitrile hydrochloride
InChI Formula InChI=1S/C27H38N2O4.ClH/c1-20(2)27(19-28,22-10-12-24(31-5)26(18-22)33-7)14-8-15-29(3)16-13-21-9-11-23(30-4)25(17-21)32-6;/h9-12,17-18,20H,8,13-16H2,1-7H3;1H/i3D3;
SMILES Cl.[2H]C([2H])([2H])N(CCCC(C#N)(C(C)C)c1ccc(OC)c(OC)c1)CCc2ccc(OC)c(OC)c2
IUPAC Name 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-(trideuteriomethyl)amino]-2-propan-2-ylpentanenitrile;hydrochloride
Molecular Weight (g/mol) 494.081
Formula Weight 493.279
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.