missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Veratridine

Product Code. 18792794
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18792794 10 mg 10mg
18702674 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18792794 Supplier R&D Systems™ Supplier No. 2918/10
  • ONLINE EXCLUSIVE PRICE
    170.86 €
    153.30 € / 10mg
    Save 17.56 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Voltage-gated Na+ channel opener

Voltage-gated Na+ channel opener; increases intracellular Ca2+ with no effect on the Na+/Ca2+ exchanger. Steroid-derived alkaloid neurotoxin. Increases membrane Na+ permeability and depolarizes excitable tissues.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-62-5
Molecular Formula C36H51NO11

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 673.79
Purity >98%
Molecular Weight (g/mol) 673.8
Chemical Name or Material Veratridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.