missing translation for 'onlineSavingsMsg'
Learn More

Verbascoside, TRC

Product Code. 30451439
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451439 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451439 Supplier Toronto Research Chemicals Supplier No. TRCV12800050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Verbascoside
CAS 61276-17-3
Purity Grade Notes HPLC
Molecular Formula C29 H36 O15
Quantity 50 mg
Synonym β-D-Glucopyranoside','2-(3,4-dihydroxyphenyl)ethyl 3-O-(6-deoxy-α-L-mannopyranosyl)-','4-[(2E)-3-(3,4-dihydroxyphenyl)-2-propenoate], β-D-Glucopyranoside','2-(3,4-dihydroxyphenyl)ethyl 3-O-(6-deoxy-α-L-mannopyranosyl)-','4-[3-(3,4-dihydroxyphenyl)-2-propenoate]','(E)-, (3,4-Dihydroxyphenyl)ethyl O-α-rhamnopyranosyl(1→3)-4-O-caffeoyl-β-D glucopyranoside, Acteoside, Distinctive Active Powder SL Special, Distinctive Phytostem Lilac, Kusaginin, NSC 603831, O'''-Desarabinosyllavandulifolioside A, Russetinol, Stereospermin, TJC 160, Verbascoside, trans-Acteoside, trans-Verbascoside
InChI Formula InChI=1S/C29H36O15/c1-13-22(36)23(37)24(38)29(41-13)44-27-25(39)28(40-9-8-15-3-6-17(32)19(34)11-15)42-20(12-30)26(27)43-21(35)7-4-14-2-5-16(31)18(33)10-14/h2-7,10-11,13,20,22-34,36-39H,8-9,12H2,1H3/b7-4+/t13-,20+,22-,23+,24+,25+,26+,27+,28+,29-/m0/s1
SMILES C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](OCCc3ccc(O)c(O)c3)O[C@H](CO)[C@H]2OC(=O)\C=C\c4ccc(O)c(O)c4)[C@H](O)[C@H](O)[C@H]1O
IUPAC Name [(2R,3R,4R,5R,6R)-6-[2-(3,4-dihydroxyphenyl)ethoxy]-5-hydroxy-2-(hydroxymethyl)-4-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-3-yl] (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
Molecular Weight (g/mol) 624.59
Formula Weight 624.2054
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.