Learn More
Linden™ Vernier Callipers with Micrometer Screw Gauge
Product Code. 13015903
Change view
Click to view available options
Length (Metric):
115 mm
150 mm
Unit Size:
Each
Material:
Plastic
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize Material
13015903 115 mm Each -
13025903 150 mm Each -
13035903 150 mm Each Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13015903 Supplier Linden™ Supplier No. H.DR39493
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Can be used for internal, external and depth measurement. Linden™ Vernier Callipers with Micrometer Screw Gauges have a micrometer screw gauge supplied in case.
  • General purpose with thumb wheel for fine adjustment
  • Metric and imperial dual graduations
  • Available in a range of lengths
Specifications
Length (Metric) 115 mm