Learn More
R&D Systems™ Mouse B7-H3 (NP_598744) VersaClone cDNA
B7-H3 has been shown to bind an unidentified counter-receptor on activated T cells to costimulate the proliferation of CD4+ or CD8+ T cells
Brand: R&D Systems™ RDC1205
Description
The Mouse B7-H3 (NP_598744) VersaClone cDNA clone contains the complete ORF for B7-H3/CD276, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The B7-H3/CD276 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Informations sur la commande
Ships at ambient temperature
Spécification
| B7-H3/CD276 | |
| 10 μg at 0.2 μg/μL | |
| 964bp | |
| B7H3, B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276, CD276 antigen, CD276 molecule, Costimulatory molecule | |
| Mouse |
| NP_598744 | |
| cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
| 80381 | |
| Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
| 10 μg |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.
For Reasearch Use Only; Not for Use in Humans