missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Versatouch™ 78-203 Coated Thermal Insulating Gloves

Product Code. 15795992 Shop All Ansell Products
Change view
Click to view available options
Size: 7
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15795992 Size: 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15795992 Supplier Ansell Edmont™ Supplier No. 78203070
  • ONLINE EXCLUSIVE PRICE
    90.60 €
    57.41 € / Pack of 12
    Save 33.19 €
    36% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offers protection from cold without sacrificing dexterity. Ansell Edmont™ Versatouch™ 78-203 Coated Thermal Insulating Gloves are light-duty gloves with a seamless knit acrylic liner that provides comfort and warmth in cold environments and has a dot-coated palm for better grip.

VersaTouch thermal insulating gloves are ideal to keep hands warm in cold environments or when working with frozen materials.
  • Micro dot coated palm for increased grip and abrasion resistance
  • Seamless knit, thermal acrylic liner provides comfort and warmth
  • Acrylic liner offers protection from cold without sacrificing dexterity
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 388/511
Coated Area Palm
Accelerator Free Yes
Applications Industrial
Coating Material PVC
Color Blue
Exterior Finish Knitted
Grade 10 Guage
Latex Free Yes
Material Acrylic
Product Type Mechanical Protection Gloves
Size 7
Sterility Non-sterile
For Use With (Application) Refrigerated/chilled areas, frozen products
Glove Cuff Style Knit
Liner Material Acrylic
Packaging Type Bag
Palm Type Micro-dot coated
Quantity 12 Pair
Length (Metric) 225 to 245 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.