missing translation for 'onlineSavingsMsg'
Learn More

Ansell VersaTouch™ 92-471 Series Light Blue Nitrile Gloves

Product Code. 12697061 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12697061 8 Pack of 100
12687061 7 Pack of 100
12607071 9 Pack of 100
12617071 10 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12697061 Supplier Ansell Supplier No. 92471080
Quantity Price
1 20.10 € / Pack of 100
10 18.38 € / Pack of 100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Made from innovative nitrile compound which offers good chemical and mechanical properties, comfort and dexterity.

  • Textured finish on fingertips
  • Improved grip
  • Contains no natural rubber
  • Suitable for contact with all fatty foods
  • Outstanding resistance to animal fats compared with PVC or natural rubber
  • Powder-free
  • No risk of contaminating food with corn starch used to powder gloves

Applications:

  • All final stages of meat or fish processing
  • Controlled-environment rooms
  • Prepared meals
  • Dairy processing
  • Catering
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Mark, Standard CATEGORY III EN 374
Product Type Controlled Environment Gloves
Size 8
Sterility Non-sterile
Thickness (Metric) 0.12 mm
Quantity 100 Pairs
Resistant To Mechanical and chemical
Series 92-471
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.