missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ VersaTouch™ 92-481 Series Light Blue Nitrile Gloves

Product Code. 12627071 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12627071 7 Pack of 100
12647071 9 Pack of 100
12637071 8 Pack of 100
12657071 10 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12627071 Supplier Ansell Edmont™ Supplier No. 92481070
Discount Lab Supplies
Use promo code 28170
View all promotions
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Made from innovative nitrile compound which offers good chemical and mechanical properties, comfort and dexterity.

  • Textured finish on fingertips
  • Improved grip
  • Contains no natural rubber
  • Suitable for contact with all fatty foods
  • Outstanding resistance to animal fats compared with PVC or natural rubber
  • Powder-free
  • No risk of contaminating food with corn starch used to powder gloves

Applications:

  • All final stages of meat or fish processing
  • Controlled-environment rooms
  • Prepared meals
  • Dairy processing
  • Catering
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Mark, Standard CATEGORY III EN 374
Accelerator Free No
Applications Industrial
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Gloves
Size 7
Sterility Non-sterile
Thickness (Metric) 0.12 mm
For Use With (Application) For all final stages of meat or fish processing, controlled-environment rooms, catering, dairy processing
Quantity 100 Pairs
Resistant To Mechanical and chemical
Series 92-481
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.