missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Versilic™ Silicone Caps

Use these stoppers for biology, microbiology and hematology, as well as food science, technical, medical and hospital apparatus. Versilic™ Silicone Caps are particularly suited for all types of tissue culture work.

Brand:  Saint Gobain Life Sciences 261020

 View more versions of this product

Product Code. 10214593

  • 270.00 € / Pack of 500
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Biologically neutral and non-toxic silicone caps for closing tubes and flasks
  • Odorless, tasteless, non-adherent and resistant to aging
  • Nontoxic, biologically neutral
  • FDA/BGA approved
  • For test tubes
  • Easy to install
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FDA Compliant Food Contact, BFR
500 Pk.
Autoclavable
20 mm
Silicone Stopper
19 mm
-70 to +250°C
10 mm
Silicone
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers