Learn More
Saint Gobain Life Sciences Versilic™ Silicone Caps
Use these stoppers for biology, microbiology and hematology, as well as food science, technical, medical and hospital apparatus. Versilic™ Silicone Caps are particularly suited for all types of tissue culture work.
Brand:  Saint Gobain Life Sciences 261014
 View more versions of this product
Product Code. 11714638
  • 825.00 € / Pack of 500
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Biologically neutral and non-toxic silicone caps for closing tubes and flasks
  • Odorless, tasteless, non-adherent and resistant to aging
  • Nontoxic, biologically neutral
  • FDA/BGA approved
  • For test tubes
  • Easy to install
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FDA Compliant Food Contact, BFR, BGA
-70 to +250°C
6 mm
Silicone
500 Pk.
Autoclavable
14 mm
Silicone Stopper