missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Verso 1-step RT-qPCR Mix, ROX

Product Code. 11872133
Change view
Click to view available options
Quantity:
200 x 25 μL Reactions
400 x 25 μL Reactions
Unit Size:
200 reactions
400 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11872133 400 x 25 μL Reactions 400 reactions
11852133 200 x 25 μL Reactions 200 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11872133 Supplier Thermo Scientific™ Supplier No. AB4101C
  • ONLINE EXCLUSIVE PRICE
    1770.00 €
    1062.00 € / 400 reactions
    Save 708.00 €
    40% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Verso RT enzyme delivers high processivity and can be used at temperatures up to 57°C, delivering effective transcription through difficult RNA secondary structures.

Thermo Scientific Verso 1-step RT-qPCR ROX Mix Kit for probe chemistry offers a quick and simple way to detect mRNA.

An optional RT Enhancer is also provided and can be used during the RT step to remove any DNA contamination, eliminating the need for DNase I treatment. All Verso 1-step RT-qPCR kits incorporate an inert dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof.

In a 1-step approach to RT-qPCR, both the RT step and qPCR steps are performed in a single tube (1-step RT-qPCR). Adopting a 1-step approach is ideal when no cDNA storage is necessary for subsequent experiments. The approach minimizes the risk of contamination and improves reproducibility by reducing sample handling.

  • Eliminate the need for DNase treatments - RT Enhancer (optional) prevents gDNA carry-over
  • RT and qPCR steps performed in single tube - convenience and reduced contamination risk
  • Use of the entire RT reaction as template for qPCR - improved sensitivity
  • Verso RT enzyme is highly sensitive with broad dynamic range - accurate detection of RNA from 1pg to 1μg
  • Verso RT enzyme has a wide working temperature range (42° to 57°C) - improved success with GC-rich and other difficult templates
  • Contains an inert blue dye for ease of pipetting during reaction set up
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains:
• 1 x 100 μL Verso Enzyme Mix
• 1 x 500 μL RT enhancer
• 1 x 5 mL 2X 1-Step qPCR ROX Mix
Sufficient for 400 x 25 μL reactions.

Store at -20°C
Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method 1-step RT-qPCR
For Use With (Equipment) 7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
Product Line Verso
Product Type 1-Step QRT-PCR Kit w/ROX
Quantity 400 x 25 μL Reactions
Sufficient For 400 Reactions
For Use With (Application) Gene Expression, Pathogen Detection
No. of Reactions 400 Reactions
Sample Type RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.