missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Verti-Gel Mini Tetrad PAGE System

Product Code. 15176624
Change view
Click to view available options
Includes:
100 x 100mm blotting module, caster and external stand, four notched/plain plates with 0.75mm spacer, four plate, four 12-well 0.75mm comb, four cassettes, eight pads
100 x 100mm blotting module, caster and external stand, four notched/plain plates with 1.5mm spacer, four plates, four 12-well 1.5mm combs, four cassettes, eight pads
100 x 100mm sliding clamps, caster and external stand, four notched/plain plates with 0.75mm bonded spacers, four notched plates, four 12-well 0.75mm combs
100 x 100mm sliding clamps, caster and external stand, four notched/plain plates with 1mm bonded spacers, four notched plates, four 12-well 1mm combs
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
15176624 100 x 100mm blotting module, caster and external stand, four notched/plain plates with 1.5mm spacer, four plates, four 12-well 1.5mm combs, four cassettes, eight pads Each
15136624 100 x 100mm sliding clamps, caster and external stand, four notched/plain plates with 0.75mm bonded spacers, four notched plates, four 12-well 0.75mm combs Each
15156624 100 x 100mm blotting module, caster and external stand, four notched/plain plates with 0.75mm spacer, four plate, four 12-well 0.75mm comb, four cassettes, eight pads Each
15116624 100 x 100mm sliding clamps, caster and external stand, four notched/plain plates with 1mm bonded spacers, four notched plates, four 12-well 1mm combs Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15176624
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 28-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Durable and leakproof system to run up to four gels.

  • Injection-moulded construction
  • Compatible with all 80 x 100mm and 100 x 100mm precast gels
  • Low buffer volumes

Specifications

Type Verti-Gel Mini Tetrad PAGE System
Gel Dimensions 75 x 80 mm
Includes 100 x 100mm blotting module, caster and external stand, four notched/plain plates with 1.5mm spacer, four plates, four 12-well 1.5mm combs, four cassettes, eight pads
Material Injection-molded
No. of Gels 4
Thickness (Metric) 0.5 mm, 0.75 mm, 1 mm, 1.5 mm, 2 mm combs
Volume (Metric) Buffer 250 to 1200 mL
Dimensions (L x W x H) 190 x 130 x 150 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.