missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Verti-Gel™ Mini Tetrad PAGE system
Shop All Fisherbrand Products
Click to view available options
Thickness (Metric) Spacer:
1 mm spacer
1.5 mm bonded spacer
Unit Size:
Each
Specifications
Specifications
| Description | With blotting module 100 x 100mm, 1mm spacer, 1mm comb |
| Device Holds | 80 Samples, 20 Samples per Gel |
| Type | Verti-Gel Mini Tetrad PAGE System |
| Includes | Blotting module 100 x 100mm, caster and external stand, four notched/plain plates with 1mm spacer, four plates, four 12-well 1mm comb, four cassette, eight pads |
| Material | Injection-molded |
| No. of Gels | 4 |
| Thickness (Metric) Spacer | 1 mm spacer |
| Volume (Metric) Buffer | 250 to 1200 mL |
| Dimensions (L x W x H) | 190 x 130 x 150 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction