missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Verti-Gel™ Mini Tetrad PAGE system

Product Code. 15146624 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Thickness (Metric) Spacer:
1 mm spacer
1.5 mm bonded spacer
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) Spacer Unit Size
15146624 1.5 mm bonded spacer Each
15126624 1 mm spacer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15146624
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

Durable and leakproof system with capacity to run up to four gels.

  • Injection-moulded construction
  • COmpatible with 80 x 100mm and 100 x 100mm precast gels
  • Low buffer volumes
  • Interchangable modules for IEF/2D electrophoresis and electroblotting in universal tank

Specifications

Description With sliding clamps 100 x 100mm, 1.5mm bonded spacer, four 12-well 1.5mm combs
Device Holds 80 Samples, 20 Samples per Gel
Type Verti-Gel Mini Tetrad PAGE System
Includes Sliding clamps 100 x 100mm,caster and external stand, 4x notched/plain plates with 1.5mm bonded spacers, four notched plates, four 12-well 1.5mm combs
Material Injection-molded
No. of Gels 4
Thickness (Metric) Spacer 1.5 mm bonded spacer
Volume (Metric) Buffer 250 to 1200 mL
Dimensions (L x W x H) 190 x 130 x 150 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.