missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Verti-Gel Maxi Dual Electroblotting System

Product Code. 15126644
Change view
Click to view available options
Device Holds:
48 Samples per Gel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Device Holds Unit Size
15126644 48 Samples per Gel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15126644 Supplier Fisherbrand™ Supplier No. FVS20CBS
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Simple-to-use casting compatible with 200 x 200mm plates.

  • Injection-moulded construction
  • Durable and leakproof
  • Rapid set-up and cooling

Specifications

Device Holds 48 Samples per Gel
For Use With (Equipment) 200 x 200 mm Plates
Type Maxi Dual Electroblotting System
Volume (Metric) Buffer 640 to 5300 mL
Dimensions (L x W x H) 300 x 180 x 270 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.