missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Vertigo Grey PU3 Gloves

Product Code. 15615947
Change view
Click to view available options
Size:
6
7
10
11
Unit Size:
10 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15615947 7 10 pairs
15605947 6 10 pairs
15645947 10 10 pairs
15655947 11 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15615947 Supplier Honeywell Safety Products™ Supplier No. 231876707
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Protect your hands when handling sharp objects in a dry or slightly greasy environment. Honeywell Safety Products™ Vertigo Grey PU3 Gloves are knitted to provide a conforming, snug fit and good dexterity.

  • Knitted glove for snug conformation to the hand.
  • Spectra™ Lycra™ thread construction for comfort, cut resistance, and dirt resistance.
  • Polyurethane-coated palm for abrasion resistance.
  • Good dexterity.
  • Ventilated back for breathability.
  • Sturdy.
  • Not for heat exposure.
  • Large cut resistance value marking (3) on the glove back for easy identification.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standard, EN 388-2003 Mechanical risks, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number 0075/797/162/11/08/0443 EXT 02/10/2015, EC Attestation Number 0075/797/162/11/08/0443 EXT 02/10/2015
Classification Dexterity: 5
Coated Area Palm, Fingers
Description Gauge 13
Coating Material Polyurethane
Color White, Gray
Cut Resistance Rating 3
Material Lycra, Spectra Fiber
Product Type Gloves
Size 7
For Use With (Application) Automotive, Aviation, Building, Construction, Catering, Energy, Electricity, Glass, Logistics, Maintenance, Metal, Paper, Service, Telecom, Transportation and Utility industries.
Puncture Rating 2
Quantity 20/PK
Tear Rating 4
Length (Metric) 23.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.