missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ VetMAX™ Xeno™ Internal Positive Control RNA

Product Code. 30057786 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Reactions:
100
500
Unit Size:
100 reactions
500 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
30057786 500 500 reactions
30057775 100 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30057786 Supplier Applied Biosystems™ Supplier No. A29761
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

VetMAX™ Xeno™ Internal Positive Control (IPC) RNA serves as an internal positive control for the RNA purification process and helps monitor for the presence of PCR inhibitors in animal health molecular detection workflows. This is of particular importance when working with sample types that have a high level of inhibitors such as oral fluids.

Xeno IPC RNA is introduced at the nucleic acid isolation/preparation step and carried through the animal health PCR workflow.

Features of VetMAX Xeno IPC RNA include:

  • Provides confidence that PCR test results are accurate and actionable
  • Easily integrates into any workflow
  • Greatly reduces the likelihood of false negatives

VetMAX Xeno IPC RNA comes in a concentration of 10,000 copies/μL and easily integrates into animal health PCR workflows, regardless of the target assay, master mix, or sample preparation reagents already in place. Coupled with a Xeno internal positive control assay, our proprietary design offers a verification layer to help ensure the qPCR test results are accurate and actionable by greatly reducing the likelihood of false negatives.

Xeno IPC RNA is a component of our licensed VetMAX Gold detection kits, is recommended in the American Association of Veterinary Diagnosticians guidelines, and has been successfully benchmarked against millions of genomes including those relevant to animal health. Xeno IPC DNA is also available (Cat. No. A29762).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage
  • 2 x 1250 μL Nucleic Acid Dilution Solution. Store at -5°C to -30°C.
  • 1 x 1250 μL Xeno RNA Control (10,000 copies/μL). Store at -5°C to -30°C.
Format Kit
For Use With (Application) Detection and Identification of Animal Disease
Includes Internal Positive Control
No. of Reactions 500
Product Line VetMAX, Xeno
Product Type Internal Positive Control RNA
Quantity 500 reactions
Shipping Condition Dry Ice
Species Not Species-Specific

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.