missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ VetMAX™ Xeno™ Internal Positive Control RNA

Product Code. p-7180678 Shop All Applied Biosystems Products
Click to view available options
No. of Reactions:
100
500
Unit Size:
100 reactions
500 reactions
This item is not returnable. View return policy

Product Code. 30057786

Brand: Applied Biosystems™ A29761

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

VetMAX™ Xeno™ Internal Positive Control (IPC) RNA serves as an internal positive control for the RNA purification process and helps monitor for the presence of PCR inhibitors in animal health molecular detection workflows. This is of particular importance when working with sample types that have a high level of inhibitors such as oral fluids.

Xeno IPC RNA is introduced at the nucleic acid isolation/preparation step and carried through the animal health PCR workflow.

Features of VetMAX Xeno IPC RNA include:

  • Provides confidence that PCR test results are accurate and actionable
  • Easily integrates into any workflow
  • Greatly reduces the likelihood of false negatives

VetMAX Xeno IPC RNA comes in a concentration of 10,000 copies/μL and easily integrates into animal health PCR workflows, regardless of the target assay, master mix, or sample preparation reagents already in place. Coupled with a Xeno internal positive control assay, our proprietary design offers a verification layer to help ensure the qPCR test results are accurate and actionable by greatly reducing the likelihood of false negatives.

Xeno IPC RNA is a component of our licensed VetMAX Gold detection kits, is recommended in the American Association of Veterinary Diagnosticians guidelines, and has been successfully benchmarked against millions of genomes including those relevant to animal health. Xeno IPC DNA is also available (Cat. No. A29762).

TRUSTED_SUSTAINABILITY

Specifikationer

Content And Storage
  • 2 x 1250 μL Nucleic Acid Dilution Solution. Store at -5°C to -30°C.
  • 1 x 1250 μL Xeno RNA Control (10,000 copies/μL). Store at -5°C to -30°C.
Format Kit
For Use With (Application) Detection and Identification of Animal Disease
Includes Internal Positive Control
No. of Reactions 500
Product Line VetMAX, Xeno
Product Type Internal Positive Control RNA
Quantity 500 reactions
Shipping Condition Dry Ice
Species Not Species-Specific

For Research Use Only. Not for use in diagnostic procedures.

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.