missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VH 032

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
VH 032 is an inhibitor of E3 ubiquitin ligase VHL (Kd = 185 nM). Blocks interaction between VHL and HIF-α downstream of HIF-α hydroxylation, initiating hypoxic response. VH 032 treatment causes dose dependent upregulation of HIF-target genes including CA9, GLUT1 and PHD2 in different cell lines. Cell permeable.
Specifications
Specifications
| Chemical Name or Material | (2 S,4 R)-1-(-2-Acetamido-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide |
| CAS | 1448188-62-2 |
| Quantity | 10 mg |
| Target | Von Hippel Lindau |
| Molecular Formula | C24H32N4O4S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction