missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VH 032 amide-alkylC5-azide
GREENER_CHOICE

Product Code. 30000018
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30000018

Brand: Tocris Bioscience™ 7867/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHL ligand with alkylC5 linker and terminal azide for onward chemistry

VH 032 amide-alkylC5-azide is a functionalized von-Hippel-Lindau (VHL) protein ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkylC5 linker and terminal azide ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.

Please contact us for SD files of our available Degrader Building Blocks.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S,4R)-1-((S)-2-(6-Azidohexanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide
CAS 2821804-11-7
Quantity 25 mg
Target Degrader Building Blocks
Molecular Formula C28H39N7O4S
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.