Learn More
VH 032 - linker 1, Tocris Biosciences™
PROTACs (Proteolysis Targeting Chimeras)
Brand: Tocris Bioscience 6463/25
Description
VH 032 amide-PEG3-amine is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand plus a PEG linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.This product has been recently renamed. The previous name for this product was VH 032 - linker 1
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
| PROTAC Toolbox | |
| 2097971-11-2 | |
| VH 032 - linker 1 | |
| 25 mg |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.