missing translation for 'onlineSavingsMsg'
Learn More

Tocris™ VH 032 - linker 6

Product Code. 18708276
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 18708276

Brand: Tocris™ 6684/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VH 032-PEG2-alkyne

VH 032 amide-PEG2-alkyne is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand plus PEG2 linker with terminal alkyne ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.

This product has been recently renamed. The previous name for this product was VH 032 - linker 6

Please contact us for SD files of our available Degrader Building Blocks.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S,4R)-1-((S)-2-(tert-Butyl)-4-oxo-6,9,12-trioxa-3-azapentadec-14-yn-1-oyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide
CAS 2098799-80-3
Synonym VH 032 - linker 6
Quantity 25 mg
Molecular Formula C31H42N4O7S
Formula Weight 614.75
Product Type Functionalized von-Hippel-Lindau (VHL) Ligand
Purity 95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.