missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VH 032, phenol
GREENER_CHOICE

Product Code. 18796196
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 18796196

Brand: Tocris Bioscience™ 6911/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydroxyl functionalized VHL ligand

VH 032, phenol is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand with terminal hydroxyl ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.

Please contact us for SD files of our available Degrader Building Blocks.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2 S,4 R)-1-(-2-acetamido-3,3-dimethylbutanoyl)-4-hydroxy-N-(2-hydroxy-4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide
CAS 2244684-42-0
Quantity 50 mg
Target Degrader Building Blocks
Molecular Formula C24H32N4O5S
Formula Weight 6666
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.