missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Vial Holder for HG-600 Geno/Grinder™ 2010 Homogenizer

This Vial Holder for HG-600 Geno/Grinder™ 2010 Homogenizer securely hold vials in place while processing samples.

Brand:  Cole-Parmer™ 2197

Code : A0

Additional Details : Weight : 2.00000kg

 View more versions of this product

Product Code. 12517336

Quantity Price
1 79.80 € / Each
10 63.28 € / Each
Estimated Shipment: 16-05-2024
to see stock.
Add to basket


Description

Description

  • Safely grind samples in the HG-600 Geno/Grinder™ 2010.
  • Vial holders are suitable for use with the HG-600 Geno/Grinder™ 2010 Homogenizer.
  • Order homogenizer and vials separately.
Specifications

Specifications

Holder
24 Standard 15 mL Conical Bottom Centrifuge Tubes
Foam
HG-600 Geno/Grinder™ 2010 Homogenizer
11.9 cm
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers