missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ ViaLight™ Plus Cell Proliferation and Cytotoxicity BioAssay Kit, 1000 Test
Descripción
- Robust - Stable bioluminescent signal (half-life >5 hours) facilitates manual or batch processing. And, unlike MTT or Alamar Blue assays, ViaLight™ Bioassays are not prone to interference due to culture condition changes.
- Flexible - Dynamic range of 5 decades and excellent performance with both adherent or suspension cultures allows you to design the experiment for your needs.
- Convenient - Simply add two reagents directly to your culture well and read. No cell washing or medium removal required.
- Sensitive - Detect as few as 10 cells allowing for lower seeding densities and more assays.
- Fast - Results from a 96-well plate can be processed and analyzed in as little as 10 minutes.
- Uses ATP bioluminescence and is rapid, sensitive and highly reproducible.
- Designed to provide unprecedented speed and sensitivity for cytotoxicity and cell proliferation studies.
- The ViaLight™ Assay incorporates bioluminescent detection of cellular ATP as a measure of viability.
- Measurement of ATP is the most accurate, effective, and direct way of determining the number of living cells in culture. The ViaLight™ Plus BioAssay is designed to deliver a high, stable luminescent signals for an extended period of time for greater experimental design flexibility.
- The easy two-step, non-radioactive assay is scalable for high-throughput applications in both 96-well and 384-well formats.
- The assay can be run on a variety of luminometers.
Especificaciones
Especificaciones
| Description | Sufficient for 1000 tests/10 plates |
| Detection Method | ATP Bioluminescence |
| For Use With (Application) | Cell proliferation, cytotoxicity |
| Includes | 2 x LT27-212 ATP monitoring reagent plus, lyophilized, 2 x LT27-079 assay buffer, 50 mL bottles, 1 x LT27-076 cell lysis reagent, 50 mL bottle |
| Quantity | 1000 Tests |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?