missing translation for 'onlineSavingsMsg'
Learn More

Nexcelom Bioscience ViaStain™ AOPI Staining Solution

Product Code. 13366169
Encompass
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13366169 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13366169 Supplier Nexcelom Bioscience Supplier No. CS201065ML

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A staining solution for the detection of live and dead nucleated mammalian cells

  • One 5 mL vial, good for approximately 250 tests
  • Product warranty is valid until expiration date stated on the product label. If no expiration date is listed, warranty is valid for 12 months from the date of receipt of the product.
  • Compatible with Cellometer Auto 2000, Auto X4, X2, K2, Vision CBA and Celigo

Specifications

For Use With (Equipment) Cellometer Auto 2000, Auto X4, X2, K2, Vision CBA and Celigo
Quantity 5 mL
compliance-icons compliance-icons
Product Identifier
  • Acridine Orange Propidium Iodide (AOPI)
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: propidium iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.