missing translation for 'onlineSavingsMsg'
Learn More

VICI Jour Chromatography™ Striped PEEK Tubing, 1/16 in.

Color-coding identifies the ID of your PEEK tubing – each ID is coded with a different color.

Brand:  VICI Jour Chromatography™ 724JRT60031M3

Code : 15

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 13123573

  • 98.25 € / 3m
Estimated Shipment: 05-06-2024
to see stock.
Add to basket

Description

Description

  • PEEK tubing has the strength required to withstand continuous use at HPLC pressure without swelling or bursting
  • Not affected by halide salts, high strength buffers, or other aggressive mobile phases that corrode stainless steel
  • Polymer surface will not leach metal ions into the eluent or extract metal-sensitive components from the sample
  • Dichloromethane, THF, and DMSO may cause swelling in PEEK, and concentrated nitric and sulphuric acid will attack PEEK
Specifications

Specifications

Tubing
Maximum Chemical Resistance and Biocompatibility.
1.58 mm
100°C
Gray
1 mm
PEEK
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers