missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VICI Jour Chromatography™ Striped PEEK Tubing, 1/16 in.
Color-coding identifies the ID of your PEEK tubing – each ID is coded with a different color.
POA - 102.00 €
Specifications
Type | Tubing |
---|---|
For Use With (Application) | Maximum Chemical Resistance and Biocompatibility. |
Diameter (Metric) Outer | 1.58 mm |
Material | PEEK |
Max. Operating Temperature (Metric) | 100°C |
Product Code | Brand | Diameter (Metric) Inner | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Color | Price | Quantity & Availability | ||||
13183563
|
VICI Jour Chromatography™
724JRT6000M3 |
0.18 mm | Yellow |
98.25 €
3m |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
13103573
|
VICI Jour Chromatography™
724JRT6002M3 |
0.5 mm | Orange |
102.00 €
3m |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
13123573
|
VICI Jour Chromatography™
724JRT60031M3 |
1 mm | Gray |
98.25 €
3m |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
13193563
|
VICI Jour Chromatography™
724JRT6001M3 |
0.25 mm | Blue |
POA
3m |
N/A | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
13133573
|
VICI Jour Chromatography™
724JRT60032M3 |
1.40 mm | Black |
98.25 €
3m |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
Description
- PEEK tubing has the strength required to withstand continuous use at HPLC pressure without swelling or bursting
- Not affected by halide salts, high strength buffers, or other aggressive mobile phases that corrode stainless steel
- Polymer surface will not leach metal ions into the eluent or extract metal-sensitive components from the sample
- Dichloromethane, THF, and DMSO may cause swelling in PEEK, and concentrated nitric and sulphuric acid will attack PEEK
Specifications
Tubing | |
1.58 mm | |
100°C |
Maximum Chemical Resistance and Biocompatibility. | |
PEEK |