Learn More
VICI Jour Chromatography™ Tubing Clip
Click to view available options
:
For holding both 1/6 in. and 1/8 in. polymer tubes in correct placement in beakers, flasks, bottles and other labe equipment up to 4mm wall thickness with stainless steel spring
Specifications
Specifications
| For Use With (Application) | holding both 1/6 in. and 1/8 in. polymer tubes in correct placement in beakers, flasks, bottles and other labe equipment up to 4mm wall thickness with stainless steel spring |
| Product Type | Tubing Clip |