Learn More
Metrohm For use with Metrohm KF Titrators.
Product Code. 10391245
Encompass_Preferred
Click to view available options
Type:
Titration Vessel (90 mL)
Unit Size:
Each
Product Code. 10391245
Brand: Metrohm 61415220
This item requires a price update prior to being ordered.
Request a Price for this item here.
  • Accessories and replacement parts
Specifications
Diameter (English) 3.07 in.
Diameter (Metric) 78 mm
Type Titration Vessel (90 mL)
Height (English) 3.14 in.
Height (Metric) 80 mm
Material Clear Glass
Volume (Metric) 20 to 90 mL