Learn More
SI Analytics™ Connection Cables and Plugs
Product Code. 10559371 Shop All SI Analytics Products
Change view
Click to view available options
Length (Metric) Cable:
1 m
2 m
5 m
10 m
Type:
K 1 A
K 10 A
K2X
K5A
L 1 R
L 2 A
Plug Cable Combination
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Cable Type unitSize
10559371 10 m K5A Each
10535553 5 m K 10 A Each
10406812 1 m K 1 A Each
10305111 1 m L 1 R Each
10466165 2 m L 2 A Each
10652832 5 m Plug Cable Combination Each
10152071 2 m K2X Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10559371 Supplier SI Analytics™ Supplier No. 285120258
Only null left
Add to basket
Add to basket
SI Analytics™ Connection Cables and Plugs are available in various combinations of electrode socket, instrument plug and cable length.
  • Coaxial plug for pH, redox, ammonia and sodium combination electrodes, pH and redox single electrodes as well as reference electrodes in Plus series. The L and LB series plugs are compatible. The LB sockets are matching colors with the BlueLine electrodes
  • Electrode plug for reference electrodes from the predecessor series
  • Plug for conductivity measuring cells with temperature sensor and oxygen cells, for older models
TRUSTED_SUSTAINABILITY
Specifications
Connector Types DIN
Type K5A
Includes 5m Coaxial Cable, DIN Instrument Plug
Length (Metric) Cable 10 m