Learn More
Poulten Selfe™ U-Tube Viscometer
Product Code. 13161023
Change view
Click to view available options
Viscosity Range:
0.9 to 3 cSt
2 to 10 cSt
20 to 100 cSt
200 to 1000 cSt
2000 to 10,000 cSt
6 to 30 cSt
60 to 300 cSt
600 to 3000 cSt
Size:
A
B
C
D
E
F
G
H
missing translation for 'unitSize'
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Range Size unitSize
13161023 0.9 to 3 cSt A Each
13171023 2 to 10 cSt B Each
13101033 60 to 300 cSt E Each
13121033 600 to 3000 cSt G Each
13131033 2000 to 10,000 cSt H Each
13191023 20 to 100 cSt D Each
13181023 6 to 30 cSt C Each
13111033 200 to 1000 cSt F Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 missing translation for 'options'
Product Code. 13161023 missing translation for 'mfr' Poulten Selfe™ missing translation for 'supplierNo' 1619/01
Suitable for transparent liquids. With UKAS certificate of calibration
  • PSL calibrated viscometers are supplied with UKAS certificates providing an official assurance of the competence of the calibration work carried out. UKAS certificates of calibration, which satisfy the requirements of ISO 9000, are issued by the PSL Calibration Laboratory, which is a National Measurement Accreditation Service accredited laboratory.
  • Type BS/U, IP71.Suitable for transparent liquids. Nominal overall length 300mm. Approx. sample size 13mL to 40mL.
Specifications
Product Type U-Tube Viscometer
Viscosity Range 0.9 to 3 cSt
Certifications/Compliance UKAS, ISO 9000
Size A
Volume (Metric) 18 mL