missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ViewRNA™ HC Screening Assay

Product Code. 32045334 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 Plates
100 Plates
50 Plates
Product Type:
HC Screen. Signal Amp., 1 Plex
HC Screen. Signal Amp., 2 Plex
HC Screen. Signal Amp., 3 Plex
HC Screen. Signal Amp., 4 Plex
HC Screening Assay Kit
Unit Size:
Pack of 10
Pack of 100
Pack of 50
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type Unit Size
32045334 10 Plates HC Screening Assay Kit Pack of 10
31722729 50 Plates HC Screening Assay Kit Pack of 50
30564884 100 Plates HC Screening Assay Kit Pack of 100
31722895 10 Plates HC Screen. Signal Amp., 1 Plex Pack of 10
31722916 50 Plates HC Screen. Signal Amp., 1 Plex Pack of 50
31722995 100 Plates HC Screen. Signal Amp., 1 Plex Pack of 100
31722954 10 Plates HC Screen. Signal Amp., 2 Plex Pack of 10
31722917 50 Plates HC Screen. Signal Amp., 2 Plex Pack of 50
31722727 100 Plates HC Screen. Signal Amp., 2 Plex Pack of 100
30565022 10 Plates HC Screen. Signal Amp., 3 Plex -
31722731 50 Plates HC Screen. Signal Amp., 3 Plex Pack of 50
31722778 100 Plates HC Screen. Signal Amp., 3 Plex Pack of 100
31722856 10 Plates HC Screen. Signal Amp., 4 Plex Pack of 10
31722921 50 Plates HC Screen. Signal Amp., 4 Plex Pack of 50
31722959 100 Plates HC Screen. Signal Amp., 4 Plex Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 32045334 Supplier Thermo Scientific™ Supplier No. QVP0011
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The ViewRNA HC Screening assay is an in situ hybridization method for visualization of up to 4 target RNAs, at single transcript detection level, simultaneously within individual cells. Dual detection with protein also possible, by optimizing experimental conditions.

The ViewRNA HC Screening Assay is an in situ hybridization method for visualization of up to four target RNAs at a single transcript detection level simultaneously within individual cells. Dual detection with protein is also possible by optimizing experimental conditions. For flexibility, the ViewRNA HC Screening Assay is provided in three modules (each sold separately): ViewRNA HC Screening Assay Kit, ViewRNA HC Screening Signal Amplification Kit, and ViewRNA Probe Sets. All three modules are needed to perform the ViewRNA HC Screening Assay. Note: the ViewRNA HC Screening Assay Kit and ViewRNA HC Screening Signal Amplification Kit are made-to-order kits. Please allow 4-6 weeks for shipment after placing order. 

Features of ViewRNA Tissue assays include:

  • Detect up to four RNA targets at single transcription level in cell culture cells in an automated format for 96- or 384-well plates
  • Utilize bright Alexa Fluor 488, Alexa Fluor 546, Alexa Fluor 947, and/or Alexa Fluor 750
  • Dual detection with proteins possible by optimizing experimental conditions

The ViewRNA assay is a novel RNA in situ hybridization solution, based on patent-pending Probe Set design and proprietary signal amplification technology. ViewRNA assays offers single copy RNA sensitivity in individual cells in a multiplex assay format. Signal amplification is predicated on specific hybridization of adjacent Probe Set oligonucleotides to a target RNA, resulting in excellent signal-to-noise ratios. Independent but compatible signal amplification systems enable simultaneous detection of up to four target RNAs in a single assay.

The ViewRNA HC Screening Assay is comprised of three modules (each sold separately):

  • ViewRNA HC Screening Assay Kit
  • ViewRNA HC Screening Signal Amplification Kit
  • ViewRNA Probe Sets. Probes can be ordered from https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cellular-imaging/in-situ-hybridization-ish/rna-fish/viewrna-cell-assays.html

How it works

Step 1: Prepare Sample. Adherent cells on a solid surface are fixed and permeabilized. Optional perform protein detection, by following immunocytochemistry protocol.

Step 2: Hybridize Probe Sets. Gene-specific Probe Sets hybridize to target mRNAs.

Step 3: Amplify Signal. Independent but compatible signal amplification systems enable simultaneous detection of multiple RNAs in a single assay. Distinct sets of PreAmp, Amp, and Label Probe molecules are used to detect different target RNAs.

Step 4: Image. Target mRNAs are visualized using high content imaging systems or standard epifluorescence microscope.

Components of ViewRNA HC Screening Assay Kits.

Store following components at 2°C to 8°C

  • Pretease QS: Enzyme in aqueous buffered solution
  • Probe Set Diluent QS: Aqueous solution containing formamide and detergent
  • Amplifier Diluent QS Aqueous solution containing formamide and detergent
  • Label Probe Diluent QS Aqueous solution containing detergent

Store following components at room temperature.

  • Protease Stop Buffer Aqueous buffered solution
  • Storage Buffer Aqueous buffered solution
  • Detergent Solution: Aqueous buffered solution containing detergent
  • Wash Buffer Component 1 (Wash Comp 1): Aqueous solution containing detergent
  • Wash Buffer Component 2 (Wash Comp 2): Aqueous buffered solution
  • Pre-Hybridization Buffer (PreHyb Buffer): Aqueous buffered solution

Components of ViewRNA HC Screening Signal Amplification Kits:

Store following components at -20°C.

  • PreAmplifier (PreAmp type 4, 6, 8, or 10): DNA in aqueous buffered solution.
  • Amplifier (Amp type 4, 6, 8, or 10): DNA in aqueous buffered solution.
  • Label Probe (LP type 4-Alexa Fluor 488, LP type 1-Alexa Fluor 546, LP type 6-Alexa Fluor 647, LP type 10-Alexa Fluor 750): Fluorescent dye-conjugated oligonucleotides in aqueous buffered solution.

Note:

  • 1-plex kits only have type 1 reagents
  • 2-plex kits have type 1 and 6 reagents
  • 3-plex kits have type 1, 4 and 6 reagents
  • 4-plex kits have type 1, 4, 6 and 10 reagents
TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorescence
Format Kit
Probe Labeled DNA, Labeled RNA
Product Line ViewRNA
Product Type HC Screening Assay Kit
Quantity 10 Plates
Target Molecule RNA (General)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.