Learn More
ViewRNA™ ISH Cell Assay Kit
Product Code. 30565021
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
Product Code. Quantity Unit Size
30565021 1 kit Each
1 options
Product Code. 30565021 Supplier Invitrogen Supplier No. QVC0001
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Content And Storage Components sufficient for 1-plex to 3-plex (AlexaFluor 488, 546 and 647) assay in 24- well plate format (24 samples), 96 assays when using cover slips mounted on glass slides
Quantity 1 kit
Type Detection Assay Kit