missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIGR/GPR126 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00 € - 468.00 €
Specifications
| Antigen | VIGR/GPR126 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
VIGR/GPR126 Polyclonal antibody specifically detects VIGR/GPR126 in Mouse samples. It is validated for Western BlotSpecifications
| VIGR/GPR126 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| PBS (pH 7.3), 50% glycerol | |
| 57211 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| APG1, Developmentally regulated G-protein-coupled receptor, DREG, FLJ14937, G protein-coupled receptor 126, G-protein coupled receptor 126, HBV PreS1-transactivated protein 2, Vascular inducible G protein-coupled receptor, vascular-inducible G protein-coupled receptor, VIGRPS1TP2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-300 of human GPR126 (NP_001027567.1). NCRVVLSNPSGTFTSPCYPNDYPNSQACMWTLRAPTGYIIQITFNDFDIEEAPNCIYDSLSLDNGESQTKFCGATAKGLSFNSSANEMHVSFSSDFSIQKKGFNASYIRVAVSLRNQKVILPQTSDAYQVSVAKSISIPELSAFTLCFEATKVGHEDSDWTAFSYSNASFTQLLSFGKAKSGYFLSISDSKCLLNNALPVKEKEDIFAESFEQLCLVWNNSLGSIGVNFKRNYETVPCDSTISKVIPGNGKLLLGSNQNEI | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title