missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex and QuantStudio™ 12k Flex System Block Upgrade Kit, 384 well

Product Code. 15749369 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 Upgrade, 1 x 384-well Plate
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15749369

Brand: Applied Biosystems™ 4453545

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Allows the following systems to operate using a 384-well block.

Reaction Speed: Standard 
TRUSTED_SUSTAINABILITY

Specifications

Format 384-well Plate
Content And Storage 384-well Block, Heated Cover, Plate Adaptor and Calibration Kit.
For Use With (Equipment) QuantStudio™
Capacity 1 x 384-well Plate
Product Line ViiA
Product Type System Block Upgrade Kit
Description 384-well Plate
Quantity 1 Upgrade, 1 x 384-well Plate

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.