missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex and QuantStudio™ 12k Flex System Block Upgrade Kit, Fast, 96 well
Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 upgrade
Unit Size:
Each
Description
Reaction Speed: Standard
Specifications
Specifications
| Format | 96-well Plate |
| Content And Storage | Fast 96-well Block (0.1mL), heated cover, plate adaptor and calibration kit. |
| For Use With (Equipment) | QuantStudio™ |
| Reaction Speed | Fast |
| Capacity | 1 x 96-well Plate |
| Product Line | ViiA |
| Product Type | System Block Upgrade Kit |
| Description | Fast 96-well Plate |
| Quantity | 1 upgrade |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction