missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ViiA™ 7 System Block Upgrade Kit, TaqMan™ Array

Product Code. 15850413 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 upgrade
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15850413 1 upgrade Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15850413 Supplier Applied Biosystems™ Supplier No. 4453546
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Allows the ViiA™ 7 Real-Time PCR System to operate using a TaqMan™ Array Block.

This kit allows the ViiA™ 7 Real-Time PCR System to operate using a TaqMan™ Array Block.

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Format 384-well Microfluidic Card
Content And Storage Includes TaqMan Array Block, heated cover, plate adaptor and calibration kit.
For Use With (Equipment) ViiA™ 7 System
Reaction Speed Standard
Capacity 1 Array Card
Product Line TaqMan
Product Type System Block Upgrade Kit
Quantity 1 upgrade

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.