missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Vilazodone hydrochloride
GREENER_CHOICE

Product Code. 18785299
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18785299 10 mg 10mg
18734868 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18785299 Supplier Tocris Bioscience™ Supplier No. 6202/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Low molecular weight, organic compound that may help regulate biological process.

Vilazodone hydrochloride is a potent 5-HT1A partial agonist and serotonin-selective reuptake inhibitor (SSRI) (IC50 values are 0.2 and 0.5 nM respectively). Antidepressant.
TRUSTED_SUSTAINABILITY

Specifications

CAS 163521-08-2
Quantity 10 mg
Target 5-HT1A
Molecular Formula C26H27N5O2.HCl
Formula Weight 477.99g/mol
Product Type Agonist

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.