missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Vinblastine sulfate

Product Code. 18075112
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18075112 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18075112 Supplier R&D Systems™ Supplier No. 1256/50
  • ONLINE EXCLUSIVE PRICE
    753.01 €
    679.35 € / 50mg
    Save 73.66 €
    9% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Disrupts microtubules

Anticancer agent; microtubule disrupter. Induces apoptosis in cultured hepatocytes and human lymphoma cells. Shown to inhibit autophagosome maturation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-67-9
Molecular Formula C46H60N4O13S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 909.06
Purity >95%
Molecular Weight (g/mol) 909.06
Chemical Name or Material Vinblastine sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.