missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vinblastine Sulfate, 98%

Product Code. 15415799
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415799 10 mg 10mg
15425799 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415799 Supplier Thermo Scientific Chemicals Supplier No. J63598.MA
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Anticancer agent; microtubule disrupter; Induces apoptosis | CAS: 143-67-9 | C46H60N4O13S, C46H60N4O13S | 909.06 g/mol

Anticancer agent; microtubule disrupter; Induces apoptosis.Vinblastine sulfate is therapeutically used as an antineoplastic. It is also used in the treatment of Hodgkin disease, choriocarcinoma, acute and chronic leukemias.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-67-9
Molecular Formula C46H60N4O13S
Synonym Vincaleukblastine sulfate

Specifications

Quantity 10 mg
Description White
Content And Storage 2°C to 8°C
Format Powder
For Use With (Application) Anticancer agent; microtubule disrupter; Induces apoptosis
Formula Weight 909.06g/mol
Melting Point 267°C
Solubility Soluble in water or DMSO
Product Type Signal Transduction Reagent
Form Powder
Beilstein 3659812
Assay 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.