missing translation for 'onlineSavingsMsg'
Learn More

Vincamine, MedChemExpress

Product Code. 16438566
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438566 100 mg 100mg
16402246 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16438566 Supplier Medchem Express Supplier No. HYB1021
 Restricted Item
Estimated Shipment: 30-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress Vincamine is a monoterpenoid indole alkaloid extracted from the Madagascar periwinkle.

Vincamine is a peripheral vasodilator and exerts a selective vasoregulator action on the brain microcapilar circulation. Vincamine is a GPR40 agonist and acts as a β-cell protector by ameliorating β-cell dysfunction and promoting glucose-stimulated insulin secretion (GSIS). Vincamine improves glucose homeostasis in vivo, and has the potential for the type 2 diabetes mellitus (T2DM) research.

Specifikationer

Chemical Name or Material Vincamine
CAS 1617-90-9
Color White
Purity Grade Notes Research
Molecular Formula C21H26N2O3
Quantity 100 mg
Solubility Information DMSO : 25 mg/mL (70.53 mM; Need ultrasonic)
SMILES O=C(OC)[C@@]1(O)C[C@@]2(CC)[C@@]3([H])C(N1C4=CC=CC=C54)=C5CCN3CCC2
Molecular Weight (g/mol) 354.44
Formula Weight 354.44
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Visa mer Visa mindre

Research purposes only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.