missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Vincristine sulfate

Product Code. 16414847
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16414847 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16414847 Supplier MedChemExpress Supplier No. HYN0488
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Vincristine sulfate is an antitumor vinca alkaloid which inhibits microtubule formation in mitotic spindle, resulting in an arrest of dividing cells at the metaphase stage. It binds to microtubule with a Ki of 85 nM.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
Molecular Weight (g/mol) 923.04 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 2068-78-2
Product Type Natural Products
Purity 97.83%
Solubility DMSO : 83.33 mg/mL (90.28 mM∣ultrasonic and warming and heat to 80°C) ∣H2O : 50 mg/mL (54.17 mM∣Need ultrasonic)
Synonym Leurocristine sulfate∣NSC-67574 sulfate∣22-Oxovincaleukoblastine sulfate
Target Microtubule/Tubulin∣Apoptosis
Quantity 50 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.