missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Vincristine sulfate
Click to view available options
:
200 mg
Specifications
Specifications
| Content And Storage | 4°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light) |
| Molecular Weight (g/mol) | 923.04 g/mol |
| Shipping Condition | Room temperature |
| For Use With (Application) | Cancer-programmed cell death |
| CAS | 2068-78-2 |
| Product Type | Natural Products |
| Purity | 97.83% |
| Solubility | DMSO : 83.33 mg/mL (90.28 mM∣ultrasonic and warming and heat to 80°C) ∣H2O : 50 mg/mL (54.17 mM∣Need ultrasonic) |
| Synonym | Leurocristine sulfate∣NSC-67574 sulfate∣22-Oxovincaleukoblastine sulfate |
| Target | Microtubule/Tubulin∣Apoptosis |
| Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction