missing translation for 'onlineSavingsMsg'
Learn More

Vinpocetine, MedChemExpress

Product Code. 16491656
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491656 500 mg 500mg
16446156 100 mg 100mg
16431797 50 mg 50mg
16431076 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491656 Supplier Medchem Express Supplier No. HY13295
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Vinpocetine (Ethyl apovincaminate) is a derivative of the alkaloid Vincamine that blocks voltage-gated Na+ channels.

The IC50 value of Vinpocetine on direct IKK inhibition in the cell-free system is 17.17 μM. Vinpocetine is a phosphodiesterase (PDE) inhibitor and inhibits NF-κB-dependent inflammatory responses by directly targeting IκB kinase complex (IKK), and has been widely used for the treatment of cerebrovascular disorders.

Specifications

Chemical Name or Material Vinpocetine
CAS 42971-09-5
Color White
Purity Grade Notes Research
Molecular Formula C22H26N2O2
Quantity 500 mg
Synonym Ethyl apovincaminate
Solubility Information Ethanol : 7.14 mg/mL (20.37 mM; Need ultrasonic) ∣DMSO : 6.25 mg/mL (17.83 mM; Need ultrasonic)
SMILES CCOC(C1=C[C@]2(CC)CCCN3CCC(C4=C(C=CC=C4)N51)=C5[C@]23[H])=O
Molecular Weight (g/mol) 350.45
Formula Weight 350.45
Percent Purity 99.34%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.