missing translation for 'onlineSavingsMsg'
Learn More

Vinyl acetate, 99+%, stabilized

Product Code. 10113480 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
25mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10113480 1 L Glass bottle 1 L
10254950 25 mL Glass bottle 25mL
10554071 2.5 L Glass bottle 2.5 L
11669758 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10113480 Supplier Thermo Scientific Acros Supplier No. 140840010
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 108-05-4 | C4H6O2 | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-05-4 , 123-31-9
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
InChI Key XTXRWKRVRITETP-UHFFFAOYSA-N
Synonym vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
PubChem CID 7904
ChEBI CHEBI:46916
IUPAC Name ethenyl acetate
SMILES CC(=O)OC=C

Specifications

Melting Point -93°C
Color Colorless
Density 0.9300g/mL
Boiling Point 72°C to 73°C
Flash Point -8°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3COOCH=CH2
Refractive Index 1.3940 to 1.396
Quantity 1 L
Beilstein 02,I,63
Fieser 01,1271; 12,565
Merck Index 15,10189
Specific Gravity 0.93
Solubility Information Solubility in water: 23g/L (20°C). Other solubilities: miscible with ethanol and ether,soluble in acetone,benzene and chloroform
Stabilizer 3 to 15ppm (hydroquinone)
Viscosity 0.43 mPa.s (20°C)
Formula Weight 86.09
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Vinyl acetate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Vinyl acetate, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Vinyl acetate, Hydroquinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.