missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Vinyl Continuous Tape

Product Code. 12890540
Change view
Click to view available options
Color:
Blue
Green
Orange
Red
White
Yellow
Length (Metric):
30 m
30.48 m
Width (Metric):
12.7 mm
29 mm
57 mm
101 mm
101.6 mm
Unit Size:
Each
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Length (Metric) Width (Metric) unitSize
12890540 White 30.48 m 101.6 mm Each
13274219 Blue 30 m 12.7 mm Each
13284219 Blue 30 m 29 mm Each
13204229 Blue 30 m 57 mm Each
13294219 Green 30 m 12.7 mm Each
13214229 Green 30 m 29 mm Each
13264229 Green 30 m 57 mm Each
15236076 Green 30 m 101 mm Each
13224229 Orange 30 m 101 mm Each
13244229 Red 30 m 12.7 mm Each
13234229 Red 30 m 29 mm Each
13254229 Red 30 m 57 mm Each
15266076 Red 30 m 101 mm Each
13274229 White 30 m 12.7 mm Each
13284229 White 30 m 29 mm Each
13294229 White 30 m 57 mm Each
15226716 Yellow 30 m 101 mm Each
13204239 Yellow 30 m 12.7 mm Each
13214239 Yellow 30 m 29 mm Each
13224239 Yellow 30 m 57 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
This item is not returnable. View return policy
Product Code. 12890540 Supplier Brady™ Supplier No. 120939
Stock Up Your Lab
Use promo code 28007
View all promotions
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

B-7569 Vinyl Continuous Tape is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels.

  • B-7569 is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels.

Specifications

Certifications/Compliance BS5609:1986 Section 2 and section 3
Description GlobalMark Indoor/Outdoor Vinyl Tape
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Gloss
Format Continuous
Material Vinyl
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Facility Identification, Floor marking, Hazard communication Identification, Pipe and valve marking, Safety Identification, Warehouse marking
For Use With (Equipment) GlobalMark Printer
Includes 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels
Labels per Row 1
Min. Operating Temperature (Metric) -20°C
No. per Pack 1 Roll/Box
Quantity 1 Ea.
Temperature Range (English) -4°F to +167°F
Temperature Range (Metric) -20°C to +75°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Dimensions (L x W) 30.48 m x 101.6 mm
Length (English) 100 ft.
Length (Metric) 30.48 m
Width (English) 4 in.
Width (Metric) 101.6 mm
Width (English) Printable Area 4 in.
Alternate Part Number GM Tape B-7569 White 100mm x 30m
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.