missing translation for 'onlineSavingsMsg'
Learn More

Vinyl iodide, tech. 85%

Product Code. 11343296
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11343296 1 g 1g
11353296 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343296 Supplier Thermo Scientific Alfa Aesar Supplier No. L12656.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 593-66-8 | C2H3I | 153.95 g/mol

Vinyl iodide is an important intermediate used in organic synthesis, chemical research and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinyl iodide is an important intermediate used in organic synthesis, chemical research and pharmaceuticals.

Solubility
Not miscible or difficult to mix in water.

Notes
It is sensitive to light. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-66-8
Molecular Formula C2H3I
Molecular Weight (g/mol) 153.95
MDL Number MFCD00039404
InChI Key GHXZPUGJZVBLGC-UHFFFAOYSA-N
Synonym vinyl iodide, ethene, iodo, iodoethylene, vinyl iodide, tech., 1-iodoethylene, iodoethyl, iodo-ethene, 1-iodoethyl, 1-iodoethylene #, vinyl iodide, tech
PubChem CID 68976
ChEBI CHEBI:51315
IUPAC Name iodoethene
SMILES IC=C

Specifications

Density 2.08
Boiling Point 55°C to 56°C
Refractive Index 1.53
Quantity 1 g
Beilstein 1731578
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 153.95
Percent Purity 85%
Grade Technical
Chemical Name or Material Vinyl iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.